siRNA Details
Gene Symbol: LINC00327 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038995.1 | 2, 3 | 849 | |||
GenBank mRNA | AL833146.1 | - | 2, 3 | 849 | ||
BX098743.1 | - | 2, 3 | 314 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00327 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038995.1 | 2, 3 | 849 | |||
GenBank mRNA | AL833146.1 | - | 2, 3 | 849 | ||
BX098743.1 | - | 2, 3 | 314 |