siRNA Details
Gene Symbol: CCDC144NL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC072410.1 | - | 3 | 751 | ||
BC113722.1 | - | 1 | 339 | |||
BC117395.1 | - | 1 | 339 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC144NL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC072410.1 | - | 3 | 751 | ||
BC113722.1 | - | 1 | 339 | |||
BC117395.1 | - | 1 | 339 |