siRNA Details
Gene Symbol: LOC101927051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109952.1 | 1 | 106 | |||
GenBank mRNA | AK097791.1 | - | 1 | 106 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB209522.1 | - | 1 | 74 | Chr. 22 - 37649878 - 37658345 [-] on Build GRCh38 |
