siRNA Details
Gene Symbol: HOTAIRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038366.1 | 3 | 726 | |||
NR_038367.1 | 2 | 457 | ||||
GenBank mRNA | BC031342.1 | - | 3 | 696 | ||
GQ479958.1 | - | 2 | 457 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HOTAIRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038366.1 | 3 | 726 | |||
NR_038367.1 | 2 | 457 | ||||
GenBank mRNA | BC031342.1 | - | 3 | 696 | ||
GQ479958.1 | - | 2 | 457 |