siRNA Details
Gene Symbol: STXBP5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034115.1 | 6 | 522 | |||
GenBank mRNA | AK091594.1 | - | 7 | 593 | ||
BC035182.2 | - | 4 | 252 | |||
BC044307.1 | - | 6 | 453 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: STXBP5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034115.1 | 6 | 522 | |||
GenBank mRNA | AK091594.1 | - | 7 | 593 | ||
BC035182.2 | - | 4 | 252 | |||
BC044307.1 | - | 6 | 453 |