siRNA Details
Gene Symbol: LINC01137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038842.1 | 1 | 145 | |||
GenBank mRNA | BM923085.1 | - | 1 | 91 | ||
DA727774.1 | - | 1 | 148 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038842.1 | 1 | 145 | |||
GenBank mRNA | BM923085.1 | - | 1 | 91 | ||
DA727774.1 | - | 1 | 148 |