siRNA Details
Gene Symbol: LINC01158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037883.1 | 3 | 358 | |||
NR_131233.1 | 3 | 239 | ||||
NR_131234.1 | 3 | 283 | ||||
NR_131235.1 | 3 | 330 | ||||
GenBank mRNA | AI879178.1 | - | 3 | 337 | ||
AK096498.1 | - | 3 | 340 | |||
AL036798.3 | - | 3 | 272 | |||
BF700938.1 | - | 3 | 356 | |||
CK004193.1 | - | 3 | 240 |
