siRNA Details
Gene Symbol: LINC01158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037883.1 | 4 | 647 | |||
NR_131233.1 | 4 | 528 | ||||
NR_131234.1 | 4 | 572 | ||||
NR_131235.1 | 4 | 619 | ||||
GenBank mRNA | AK096498.1 | - | 4 | 629 | ||
CK004193.1 | - | 4 | 529 |
