siRNA Details
Gene Symbol: LOC55338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130914.1 | 3 | 1111 | |||
GenBank mRNA | AK002104.1 | - | 2 | 1805 | ||
AK002154.1 | - | 2 | 1032 | |||
AK023417.1 | - | 3 | 1112 | |||
AK024354.1 | - | 1 | 758 |
