siRNA Details
Gene Symbol: LOC100506098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134567.1 | 5 | 650 | |||
NR_134568.1 | 5 | 559 | ||||
NR_134569.1 | 4 | 488 | ||||
GenBank mRNA | BG777800.1 | - | 4 | 489 | ||
BI759100.1 | - | 5 | 550 | |||
DR423379.1 | - | 5 | 635 |
