siRNA Details
Gene Symbol: LOC101927502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109772.1 | 3 | 449 | |||
GenBank mRNA | BC036431.2 | - | 3 | 449 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109772.1 | 3 | 449 | |||
GenBank mRNA | BC036431.2 | - | 3 | 449 |