siRNA Details
Gene Symbol: CTC-338M12.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109909.1 | 3 | 738 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK026811.1 | - | 1 | 1594 | Chr. 5 - 181255719 - 181257586 [+] on Build GRCh38 |
