siRNA Details
Gene Symbol: LOC101928834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120646.1 | 4 | 374 | |||
GenBank mRNA | BC014575.1 | - | 4 | 378 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE747996.1 | - | 4 | 355 | Chr. 10 - 19711028 - 19728531 [-] on Build GRCh38 |
