siRNA Details
Gene Symbol: RNASEH1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038429.1 | 1 | 439 | |||
NR_038430.1 | 1 | 438 | ||||
NR_038431.1 | 1 | 438 | ||||
GenBank mRNA | AK023018.1 | - | 1 | 277 | ||
BC040057.1 | - | 1 | 333 |
