siRNA Details
Gene Symbol: RNASEH1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038429.1 | 1 | 601 | |||
NR_038430.1 | 1 | 600 | ||||
NR_038431.1 | 1 | 600 | ||||
GenBank mRNA | BC040057.1 | - | 1 | 495 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ221780.1 | - | 1 | 493 | Chr. 2 - 3558494 - 3559676 [+] on Build GRCh38 |
