siRNA Details
Gene Symbol: LINC01114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038231.1 | 1 | 416 | |||
GenBank mRNA | AK096621.1 | - | 1 | 416 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038231.1 | 1 | 416 | |||
GenBank mRNA | AK096621.1 | - | 1 | 416 |