siRNA Details
Gene Symbol: LOC100507468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108105.1 | 2 | 388 | |||
GenBank mRNA | BX097309.1 | - | 2 | 305 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100507468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108105.1 | 2 | 388 | |||
GenBank mRNA | BX097309.1 | - | 2 | 305 |