siRNA Details
Gene Symbol: JAZF1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034097.1 | 4 | 296 | |||
GenBank mRNA | AK293540.1 | - | 4 | 296 | ||
BC043567.1 | - | 2 | 246 | |||
DB481568.1 | - | 3 | 187 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: JAZF1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034097.1 | 4 | 296 | |||
GenBank mRNA | AK293540.1 | - | 4 | 296 | ||
BC043567.1 | - | 2 | 246 | |||
DB481568.1 | - | 3 | 187 |