siRNA Details
Gene Symbol: MAGI2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038344.1 | 5 | 872 | |||
NR_038345.1 | 4 | 700 | ||||
NR_038346.1 | 4 | 683 | ||||
GenBank mRNA | AB371456.1 | - | 4 | 546 | ||
BI868593.1 | - | 4 | 482 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB543548.1 | - | 2 | 290 | Chr. 7 - 79458936 - 79471194 [+] on Build GRCh38 |
