siRNA Details
Gene Symbol: MAGI2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038343.2 | 3 | 446 | |||
NR_038344.1 | 3 | 446 | ||||
NR_038345.1 | 2 | 274 | ||||
NR_038346.1 | 3 | 343 | ||||
GenBank mRNA | AB371456.1 | - | 2 | 120 | ||
AK311103.1 | - | 3 | 414 | |||
AL833456.1 | - | 1 | 170 | |||
BI521274.1 | - | 3 | 447 | |||
BI868593.1 | - | 3 | 142 | |||
CN311982.1 | - | 3 | 413 | |||
DA758742.1 | - | 2 | 274 |
