siRNA Details
Gene Symbol: MAGI2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038343.2 | 4 | 756 | |||
GenBank mRNA | AK311103.1 | - | 4 | 724 | ||
AL833456.1 | - | 2 | 480 | |||
CR749529.1 | - | 2 | 809 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MAGI2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038343.2 | 4 | 756 | |||
GenBank mRNA | AK311103.1 | - | 4 | 724 | ||
AL833456.1 | - | 2 | 480 | |||
CR749529.1 | - | 2 | 809 |