siRNA Details
Gene Symbol: RPL34-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026968.1 | 4 | 1419 | |||
GenBank mRNA | AK094992.1 | - | 4 | 1360 | ||
AK298931.1 | - | 4 | 760 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RPL34-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026968.1 | 4 | 1419 | |||
GenBank mRNA | AK094992.1 | - | 4 | 1360 | ||
AK298931.1 | - | 4 | 760 |