siRNA Details
Gene Symbol: SP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103856.1 | 2, 3 | 191 | |||
GenBank mRNA | BM804969.1 | - | 2, 3 | 166 | ||
HY111832.1 | - | 2, 3 | 192 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI768292.1 | - | 2, 3 | 145 | Chr. 17 - 47899757 - 47941359 [-] on Build GRCh38 |
