siRNA Details
Gene Symbol: SDCBP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040047.1 | 3 | 1449 | |||
NR_040048.1 | 4 | 1411 | ||||
NR_040049.1 | 3 | 1196 | ||||
GenBank mRNA | AK055993.1 | - | 4 | 1333 | ||
BC034773.1 | - | 3 | 1129 |
