siRNA Details
Gene Symbol: SDCBP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040047.1 | 3 | 868 | |||
NR_040048.1 | 4 | 830 | ||||
NR_040049.1 | 3 | 615 | ||||
GenBank mRNA | BC034773.1 | - | 3 | 548 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI818721.1 | - | 3 | 548 | Chr. 20 - 1325410 - 1377362 [+] on Build GRCh38 |
