siRNA Details
Gene Symbol: FAM170B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038973.1 | 1 | 399 | |||
GenBank mRNA | AK093055.1 | - | 1 | 401 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM170B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038973.1 | 1 | 399 | |||
GenBank mRNA | AK093055.1 | - | 1 | 401 |