siRNA Details
Gene Symbol: FOXP4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126415.1 | 2 | 513 | |||
NR_126416.1 | 2 | 354 | ||||
NR_126417.1 | 2 | 424 | ||||
GenBank mRNA | BG028463.1 | - | 2 | 357 | ||
BQ128057.1 | - | 2 | 361 | |||
BQ581966.1 | - | 2 | 396 |
