siRNA Details
Gene Symbol: LINC01103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110113.1 | 3, 4 | 629 | |||
GenBank mRNA | BC040913.1 | - | 3, 4 | 629 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM908150.1 | - | 641 | Chr. - - [-] on Build |
