siRNA Details
Gene Symbol: SLIT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038330.1 | 1 | 205 | |||
GenBank mRNA | BC021189.2 | - | 1 | 205 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLIT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038330.1 | 1 | 205 | |||
GenBank mRNA | BC021189.2 | - | 1 | 205 |