siRNA Details
Gene Symbol: SOX9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103737.1 | 2 | 75 | |||
NR_103738.1 | 4 | 458 | ||||
GenBank mRNA | AK056229.1 | - | 5 | 1663 | ||
AK094963.1 | - | 4 | 458 | |||
AL833139.1 | - | 2 | 75 |
