siRNA Details
Gene Symbol: DPP10-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036580.1 | 2 | 267 | |||
GenBank mRNA | AB593078.1 | - | 2 | 289 | ||
BC032913.2 | - | 2 | 267 | |||
BC048425.1 | - | 2 | 260 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DPP10-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036580.1 | 2 | 267 | |||
GenBank mRNA | AB593078.1 | - | 2 | 289 | ||
BC032913.2 | - | 2 | 267 | |||
BC048425.1 | - | 2 | 260 |