siRNA Details
Gene Symbol: LINC00940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036546.1 | 2 | 793 | |||
GenBank mRNA | AK057909.1 | - | 2 | 751 | ||
AK296286.1 | - | 2 | 751 | |||
AK316036.1 | - | 2 | 791 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036546.1 | 2 | 793 | |||
GenBank mRNA | AK057909.1 | - | 2 | 751 | ||
AK296286.1 | - | 2 | 751 | |||
AK316036.1 | - | 2 | 791 |