siRNA Details
Gene Symbol: LINC00940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036546.1 | 2 | 333 | |||
GenBank mRNA | AK296286.1 | - | 2 | 291 | ||
AK316036.1 | - | 2 | 331 | |||
DA269488.1 | - | 2 | 333 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036546.1 | 2 | 333 | |||
GenBank mRNA | AK296286.1 | - | 2 | 291 | ||
AK316036.1 | - | 2 | 331 | |||
DA269488.1 | - | 2 | 333 |