siRNA Details
Gene Symbol: ZRANB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038420.1 | 3 | 1667 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG210094.1 | - | 2 | 548 | Chr. 1 - 71048853 - 71067189 [+] on Build GRCh38 |
