siRNA Details
Gene Symbol: FLJ41941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024417.1 | 2 | 193 | |||
GenBank mRNA | AK123935.1 | - | 2 | 193 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA826529.1 | - | 2 | 193 | Chr. 22 - 18029385 - 18035887 [+] on Build GRCh38 |
