siRNA Details
Gene Symbol: EPN2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_048576.1 | 3 | 731 | |||
GenBank mRNA | BC037786.2 | - | 3 | 730 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: EPN2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_048576.1 | 3 | 731 | |||
GenBank mRNA | BC037786.2 | - | 3 | 730 |