siRNA Details
Gene Symbol: EPN2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_048576.1 | 1, 2 | 317 | |||
GenBank mRNA | BC037786.2 | - | 1, 2 | 316 | ||
BG705989.1 | - | 1, 2 | 321 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: EPN2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_048576.1 | 1, 2 | 317 | |||
GenBank mRNA | BC037786.2 | - | 1, 2 | 316 | ||
BG705989.1 | - | 1, 2 | 321 |