siRNA Details
Gene Symbol: LOC100996342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103793.1 | 2 | 480 | |||
GenBank mRNA | BC021738.1 | - | 2 | 488 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100996342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103793.1 | 2 | 480 | |||
GenBank mRNA | BC021738.1 | - | 2 | 488 |