siRNA Details
Gene Symbol: DICER1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127179.1 | - | 11 | |||
BX648898.1 | - | 1 | 3731 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DICER1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127179.1 | - | 11 | |||
BX648898.1 | - | 1 | 3731 |