siRNA Details
Gene Symbol: DPYD-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046591.1 | 1 | 109 | |||
GenBank mRNA | BU569038.1 | - | 1 | 69 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DPYD-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046591.1 | 1 | 109 | |||
GenBank mRNA | BU569038.1 | - | 1 | 69 |