siRNA Details
Gene Symbol: DPYD-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046591.1 | 1 | 137 | |||
GenBank mRNA | BU569038.1 | - | 1 | 97 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU569270.1 | - | 1 | 137 | Chr. 1 - 97796921 - 97797859 [+] on Build GRCh38 |
