siRNA Details
Gene Symbol: OLMALINC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026762.1 | 1 | 258 | |||
GenBank mRNA | BC024924.1 | - | 1 | 219 | ||
BC045787.1 | - | 1 | 237 | |||
CB159470.1 | - | 1 | 285 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OLMALINC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026762.1 | 1 | 258 | |||
GenBank mRNA | BC024924.1 | - | 1 | 219 | ||
BC045787.1 | - | 1 | 237 | |||
CB159470.1 | - | 1 | 285 |