siRNA Details
Gene Symbol: MACROD2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037841.3 | 3 | 483 | |||
NR_110318.1 | 2 | 274 | ||||
GenBank mRNA | BX417920.2 | - | 3 | 461 | ||
HY015789.1 | - | 2 | 274 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MACROD2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037841.3 | 3 | 483 | |||
NR_110318.1 | 2 | 274 | ||||
GenBank mRNA | BX417920.2 | - | 3 | 461 | ||
HY015789.1 | - | 2 | 274 |