siRNA Details
Gene Symbol: NEXN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103535.1 | 3 | 2136 | |||
GenBank mRNA | AK075118.1 | - | 4 | 2117 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NEXN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103535.1 | 3 | 2136 | |||
GenBank mRNA | AK075118.1 | - | 4 | 2117 |