siRNA Details
Gene Symbol: ASMTL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026710.1 | 2 | 721 | |||
NR_026711.1 | 2 | 721 | ||||
GenBank mRNA | AF193043.2 | - | 1 | 84 | ||
AK023392.1 | - | 2 | 154 | |||
BC093994.2 | - | 1 | 64 | |||
BC143527.1 | - | 1 | 64 |
