siRNA Details
Gene Symbol: TMEM147-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038396.1 | 3, 4 | 773 | |||
GenBank mRNA | BC024262.1 | - | 2, 3 | 2447 | ||
CR627053.1 | - | 3, 4 | 773 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM147-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038396.1 | 3, 4 | 773 | |||
GenBank mRNA | BC024262.1 | - | 2, 3 | 2447 | ||
CR627053.1 | - | 3, 4 | 773 |