siRNA Details
Gene Symbol: LINC01361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110633.1 | 1 | 24 | |||
GenBank mRNA | U80773.1 | - | 1 | 24 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | R60460.1 | - | 24 | Chr. - - [-] on Build |
