siRNA Details
Gene Symbol: LINC01134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024455.1 | 3 | 445 | |||
GenBank mRNA | BC016994.1 | - | 3 | 433 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG032857.1 | - | 3 | 431 | Chr. 1 - 3900417 - 3914157 [+] on Build GRCh38 |
