siRNA Details
Gene Symbol: LOC101927683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110683.1 | 1 | 134 | |||
GenBank mRNA | AK093200.1 | - | 1 | 134 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB026555.1 | - | 1 | 134 | Chr. 1 - 232730613 - 232742715 [-] on Build GRCh38 |
