siRNA Details
Gene Symbol: DNAJC9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038373.1 | 4 | 425 | |||
NR_134458.1 | 3 | 605 | ||||
GenBank mRNA | BC043233.1 | - | 4 | 433 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI560079.1 | - | 4 | 433 | Chr. 10 - 73247360 - 73274941 [+] on Build GRCh38 |
